Product Details

SNP ID
rs6026544
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:58819365 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACTCACCTGCGGCATGATCGCCTG[C/T]GGTCACACTCCAACTTGATATCTTG
Phenotype
MIM: 610540 MIM: 610945 MIM: 614914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAS-AS1 PubMed Links

Gene Details

Gene
GNAS-AS1
Gene Name
GNAS antisense RNA 1
There are no transcripts associated with this gene.

Gene
MIR296
Gene Name
microRNA 296
There are no transcripts associated with this gene.

Gene
MIR298
Gene Name
microRNA 298
There are no transcripts associated with this gene.

View Full Product Details