Product Details

SNP ID
rs3738480
Assay Type
Functionally tested
NCBI dbSNP Submissions
82
Location
Chr.1:151018349 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTTCATTTTTATAGACTGGGTACTT[A/G]TGGCTTTGAGAAAGGTGGTAACTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM63A PubMed Links

Gene Details

Gene
FAM63A
Gene Name
family with sequence similarity 63 member A
There are no transcripts associated with this gene.

Gene
PRUNE1
Gene Name
prune exopolyphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303229.1 Intron NP_001290158.1
NM_001303242.1 Intron NP_001290171.1
NM_001303243.1 Intron NP_001290172.1
NM_021222.2 Intron NP_067045.1
XM_005245393.4 Intron XP_005245450.1
XM_011509832.2 Intron XP_011508134.1
XM_017001955.1 Intron XP_016857444.1
XM_017001956.1 Intron XP_016857445.1
XM_017001957.1 Intron XP_016857446.1

View Full Product Details