Product Details

SNP ID
rs61995731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:899208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACGGCGGGAACCCATGCAGCAC[C/T]GTGTAGCCACTCTCCTTGCTGCCAA
Phenotype
MIM: 608114 MIM: 612056
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAP1 PubMed Links

Gene Details

Gene
ADAP1
Gene Name
ArfGAP with dual PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284308.1 954 Silent Mutation ACA,ACG T318T NP_001271237.1
NM_001284309.1 954 Silent Mutation ACA,ACG T235T NP_001271238.1
NM_001284310.1 954 Silent Mutation ACA,ACG T235T NP_001271239.1
NM_001284311.1 954 Silent Mutation ACA,ACG T212T NP_001271240.1
NM_006869.3 954 Silent Mutation ACA,ACG T307T NP_006860.1
Gene
GET4
Gene Name
golgi to ER traffic protein 4
There are no transcripts associated with this gene.

View Full Product Details