Product Details
- SNP ID
-
rs34781001
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:128620588 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGTGCATCCAGCTTCTGCATTTC[A/G]CTCACCTGGCCGAGGCAAGAGCAGG
- Phenotype
-
MIM: 180470
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPN1
PubMed Links
Gene Details
- Gene
- RPN1
- Gene Name
- ribophorin I
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002950.3 |
1723 |
Silent Mutation |
AGC,AGT |
S549S |
NP_002941.1 |
View Full Product Details