Product Details

SNP ID
rs4451579
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:248323714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGAGGCTATAAATGAGAGGATT[C/T]AGCATGGGAGTGACGATGGTGTAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2M7 PubMed Links

Gene Details

Gene
OR2M7
Gene Name
olfactory receptor family 2 subfamily M member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004691.1 Intron NP_001004691.1

View Full Product Details