Product Details

SNP ID
rs17742683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43801706 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGATGTTCTGGGATAAAGTTACC[C/T]GACCCAACTAACAGGTACCCAGTGA
Phenotype
MIM: 601114
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPP3 PubMed Links

Gene Details

Gene
MPP3
Gene Name
membrane palmitoylated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001932.4 1928 Missense Mutation AGG,GGG R585G NP_001923.2
XM_006721915.2 1928 Missense Mutation AGG,GGG R585G XP_006721978.1
XM_006721916.3 1928 Intron XP_006721979.1
XM_011524828.2 1928 Missense Mutation AGG,GGG R610G XP_011523130.1
XM_017024655.1 1928 Intron XP_016880144.1
XM_017024656.1 1928 Missense Mutation AGG,GGG R383G XP_016880145.1
XM_017024657.1 1928 Missense Mutation AGG,GGG R355G XP_016880146.1
XM_017024658.1 1928 Intron XP_016880147.1
XM_017024659.1 1928 Intron XP_016880148.1
XM_017024660.1 1928 Intron XP_016880149.1

View Full Product Details