Product Details

SNP ID
rs150237752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33056517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGAGGTTGGAGTTTCTAGTAC[C/T]TTCCTACTTCCACTCTCTCTGCAGC
Phenotype
MIM: 615717
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPIFB3 PubMed Links

Gene Details

Gene
BPIFB3
Gene Name
BPI fold containing family B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182658.2 Intron NP_872599.1

View Full Product Details