Product Details

SNP ID
rs75321624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85844344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTGAGCCAAAGTGAAATTTGTA[C/T]ACATCGCCCCTCAAACAGGGAATGA
Phenotype
MIM: 604402
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST3GAL5 PubMed Links

Gene Details

Gene
ST3GAL5
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042437.1 Intron NP_001035902.1
NM_003896.3 Intron NP_003887.3
XM_005264630.4 Intron XP_005264687.1
XM_011533143.2 Intron XP_011531445.1
XM_017005199.1 Intron XP_016860688.1
XM_017005200.1 Intron XP_016860689.1
XM_017005201.1 Intron XP_016860690.1
XM_017005202.1 Intron XP_016860691.1
XM_017005203.1 Intron XP_016860692.1
XM_017005204.1 Intron XP_016860693.1
XM_017005205.1 Intron XP_016860694.1
XM_017005206.1 Intron XP_016860695.1
XM_017005207.1 Intron XP_016860696.1
XM_017005208.1 Intron XP_016860697.1
XM_017005209.1 Intron XP_016860698.1
XM_017005210.1 Intron XP_016860699.1
XM_017005211.1 Intron XP_016860700.1
XM_017005212.1 Intron XP_016860701.1
XM_017005213.1 Intron XP_016860702.1
XM_017005214.1 Intron XP_016860703.1
XM_017005215.1 Intron XP_016860704.1

View Full Product Details