Product Details

SNP ID
rs542385576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30987821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATATTTTTTGAGGGCCTAATTTT[A/G]TTTTTAAAAACTTGCTGGGAATTAA
Phenotype
MIM: 611358
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF135 PubMed Links

Gene Details

Gene
RNF135
Gene Name
ring finger protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184992.1 Intron NP_001171921.1
NM_032322.3 Intron NP_115698.3
NM_197939.1 Intron NP_922921.1
XM_005258043.4 Intron XP_005258100.1
XM_017025223.1 Intron XP_016880712.1

View Full Product Details