Product Details

SNP ID
rs2958154
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:56671929 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATAGCTTGTCATTTCTCACCTTT[C/T]TCATTTCTTTACCTAAATTTTCTCT
Phenotype
MIM: 607061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGES3 PubMed Links

Gene Details

Gene
PTGES3
Gene Name
prostaglandin E synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282601.1 Intron NP_001269530.1
NM_001282602.1 Intron NP_001269531.1
NM_001282603.1 Intron NP_001269532.1
NM_001282604.1 Intron NP_001269533.1
NM_001282605.1 Intron NP_001269534.1
NM_006601.6 Intron NP_006592.3
XM_005268576.4 Intron XP_005268633.1
XM_006719199.2 Intron XP_006719262.1
XM_011537773.2 Intron XP_011536075.1
XM_011537774.2 Intron XP_011536076.1
XM_017018716.1 Intron XP_016874205.1

View Full Product Details