Product Details

SNP ID
rs12734733
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:247814833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGAGCTTTCCCTTTATCAACAT[A/C]CCCAGAGCCACCTTTATGGCCTTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR14A16 PubMed Links

Gene Details

Gene
OR14A16
Gene Name
olfactory receptor family 14 subfamily A member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001966.1 897 Silent Mutation NP_001001966.1

View Full Product Details