Product Details

SNP ID
rs17885718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65183167 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCCAGTTCATTGTGGATGGAGCT[G/A]CCCGCACAGACATCTGCCAGGGAGC
Phenotype
MIM: 114220 MIM: 614030
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CAPN1 PubMed Links

Gene Details

Gene
CAPN1
Gene Name
calpain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198868.1 504 Missense Mutation ACC,GCC T103A NP_001185797.1
NM_001198869.1 504 Missense Mutation ACC,GCC T103A NP_001185798.1
NM_005186.3 504 Missense Mutation ACC,GCC T103A NP_005177.2
XM_006718698.1 504 Missense Mutation ACC,GCC T103A XP_006718761.1
XM_011545292.1 504 Missense Mutation ACC,GCC T103A XP_011543594.1
Gene
SPDYC
Gene Name
speedy/RINGO cell cycle regulator family member C
There are no transcripts associated with this gene.

View Full Product Details