Product Details

SNP ID
rs7970523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56598593 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAAGGTGGGGGGAGATGCCAC[A/C]AGGTGACTCCCCACCTCCCTTGCCT
Phenotype
MIM: 605682 MIM: 602387
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BAZ2A PubMed Links

Gene Details

Gene
BAZ2A
Gene Name
bromodomain adjacent to zinc finger domain 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300905.1 5831 UTR 3 NP_001287834.1
NM_013449.3 5831 UTR 3 NP_038477.2
XM_011537797.2 5831 UTR 3 XP_011536099.1
XM_011537798.2 5831 UTR 3 XP_011536100.1
XM_011537799.2 5831 UTR 3 XP_011536101.1
XM_011537800.2 5831 UTR 3 XP_011536102.1
XM_011537801.2 5831 UTR 3 XP_011536103.1
XM_011537803.2 5831 UTR 3 XP_011536105.1
XM_011537804.2 5831 UTR 3 XP_011536106.1
XM_011537805.2 5831 UTR 3 XP_011536107.1
XM_011537806.2 5831 UTR 3 XP_011536108.1
XM_011537807.2 5831 UTR 3 XP_011536109.1
XM_011537809.2 5831 UTR 3 XP_011536111.1
XM_017018740.1 5831 UTR 3 XP_016874229.1
XM_017018741.1 5831 UTR 3 XP_016874230.1
XM_017018742.1 5831 UTR 3 XP_016874231.1
Gene
RBMS2
Gene Name
RNA binding motif single stranded interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details