Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001077394.1 | Intron | NP_001070862.1 | |||
NM_001077395.1 | Intron | NP_001070863.1 | |||
NM_015958.2 | Intron | NP_057042.2 | |||
XM_005270938.1 | Intron | XP_005270995.1 | |||
XM_005270939.4 | Intron | XP_005270996.1 | |||
XM_005270941.3 | Intron | XP_005270998.1 | |||
XM_006710683.3 | Intron | XP_006710746.1 | |||
XM_011541569.1 | Intron | XP_011539871.1 | |||
XM_011541570.2 | Intron | XP_011539872.1 | |||
XM_017001460.1 | Intron | XP_016856949.1 | |||
XM_017001461.1 | Intron | XP_016856950.1 | |||
XM_017001462.1 | Intron | XP_016856951.1 |