Product Details

SNP ID
rs6540771
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:212997030 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATATATTACCAATCTGTCTATTT[C/T]GAAATATTTTTACCTTCTAGATCTA
Phenotype
MIM: 610471
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANGEL2 PubMed Links

Gene Details

Gene
ANGEL2
Gene Name
angel homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300753.1 Intron NP_001287682.1
NM_001300755.1 Intron NP_001287684.1
NM_001300757.1 Intron NP_001287686.1
NM_001300758.1 Intron NP_001287687.1
NM_144567.4 Intron NP_653168.2
XM_005273344.1 Intron XP_005273401.1
XM_005273345.1 Intron XP_005273402.1
XM_005273346.2 Intron XP_005273403.1
XM_005273347.3 Intron XP_005273404.1
XM_017002774.1 Intron XP_016858263.1
XM_017002775.1 Intron XP_016858264.1
XM_017002776.1 Intron XP_016858265.1
XM_017002777.1 Intron XP_016858266.1
XM_017002778.1 Intron XP_016858267.1
Gene
VASH2
Gene Name
vasohibin 2
There are no transcripts associated with this gene.

View Full Product Details