Product Details

SNP ID
rs3743138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40288963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCCTGGCTCAGGAGGGGCCTG[A/C]TGGCCACCCTCGCTCCCTGGACAGT
Phenotype
MIM: 604114
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD63 PubMed Links

Gene Details

Gene
ANKRD63
Gene Name
ankyrin repeat domain 63
There are no transcripts associated with this gene.

Gene
PLCB2
Gene Name
phospholipase C beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284297.1 Intron NP_001271226.1
NM_001284298.1 Intron NP_001271227.1
NM_001284299.1 Intron NP_001271228.1
NM_004573.2 Intron NP_004564.2
XM_011521674.2 Intron XP_011519976.1
XM_017022314.1 Intron XP_016877803.1
XM_017022315.1 Intron XP_016877804.1
XM_017022316.1 Intron XP_016877805.1
XM_017022317.1 Intron XP_016877806.1
XM_017022318.1 Intron XP_016877807.1
XM_017022319.1 Intron XP_016877808.1

View Full Product Details