Product Details

SNP ID
rs6904236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30628358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGAGTGGAACCGCACCAACTGGC[A/G]ATTGACCGACCCTCACAGAAGCTGC
Phenotype
MIM: 615556 MIM: 611982 MIM: 603771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATAT1 PubMed Links

Gene Details

Gene
ATAT1
Gene Name
alpha tubulin acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031722.3 509 Silent Mutation GCA,GCG A143A NP_001026892.1
NM_001190724.2 509 Silent Mutation GCA,GCG A143A NP_001177653.1
NM_001254952.2 509 Silent Mutation GCA,GCG A155A NP_001241881.1
NM_001318762.1 509 Silent Mutation GCA,GCG A155A NP_001305691.1
NM_001318763.1 509 Silent Mutation GCA,GCG A155A NP_001305692.1
NM_024909.3 509 Silent Mutation GCA,GCG A155A NP_079185.2
XM_005249415.2 509 Silent Mutation GCA,GCG A143A XP_005249472.1
XM_011514918.2 509 Intron XP_011513220.1
XM_017011316.1 509 Silent Mutation GCA,GCG A143A XP_016866805.1
Gene
MRPS18B
Gene Name
mitochondrial ribosomal protein S18B
There are no transcripts associated with this gene.

Gene
PPP1R10
Gene Name
protein phosphatase 1 regulatory subunit 10
There are no transcripts associated with this gene.

View Full Product Details