Product Details

SNP ID
rs12015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:43023729 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGACGCCGACGTCGACCAGCAG[A/G]GACTGGTGAGTCCTGCCAGCGGCCC
Phenotype
MIM: 615339
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC15 PubMed Links

Gene Details

Gene
DNAJC15
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
There are no transcripts associated with this gene.

View Full Product Details