Product Details

SNP ID
rs11550540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46687130 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGAGTTTTTTCAGTCTCCATAG[C/T]GGACTGAATATAATCTCGAACAGTC
Phenotype
MIM: 606411 MIM: 608679
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC13A3 PubMed Links

Gene Details

Gene
SLC13A3
Gene Name
solute carrier family 13 member 3
There are no transcripts associated with this gene.

Gene
TP53RK
Gene Name
TP53 regulating kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033550.3 608 Missense Mutation ACT,GCT T129A NP_291028.3

View Full Product Details