Product Details

SNP ID
rs79692641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184236119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTGTATCAGGGCCCATGGGGC[C/T]GGCCTCACGCCGCCCTCCCTGGCCC
Phenotype
MIM: 608750 MIM: 611620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG3 PubMed Links

Gene Details

Gene
ALG3
Gene Name
ALG3, alpha-1,3- mannosyltransferase
There are no transcripts associated with this gene.

Gene
MIR1224
Gene Name
microRNA 1224
There are no transcripts associated with this gene.

Gene
VWA5B2
Gene Name
von Willebrand factor A domain containing 5B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320373.1 Intron NP_001307302.1
NM_138345.2 Intron NP_612354.1
XM_011513288.2 Intron XP_011511590.1
XM_011513289.2 Intron XP_011511591.1
XM_011513290.2 Intron XP_011511592.1
XM_011513291.2 Intron XP_011511593.1
XM_011513292.2 Intron XP_011511594.1

View Full Product Details