Product Details

SNP ID
rs117055290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52124357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGAGTGGAAATGAGAGCTCGG[C/T]GAGGCTGACCAGGCCCACCTGAGCT
Phenotype
MIM: 604447
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNB5 PubMed Links

Gene Details

Gene
GNB5
Gene Name
G protein subunit beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006578.3 Intron NP_006569.1
NM_016194.3 Intron NP_057278.2
XM_011521162.2 Intron XP_011519464.1
XM_011521163.2 Intron XP_011519465.1
XM_017021867.1 Intron XP_016877356.1

View Full Product Details