Product Details
- SNP ID
-
hCV25760545
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:160343114 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGACATCCAACCTCCATGTTCCCC[C/G]TTTTATTCTCCACTACCTTTGACCC
- Phenotype
-
MIM: 601924
MIM: 605254
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
COPA
PubMed Links
Gene Details
- Gene
- COPA
- Gene Name
- coatomer protein complex subunit alpha
There are no transcripts associated with this gene.
- Gene
- NCSTN
- Gene Name
- nicastrin
There are no transcripts associated with this gene.
View Full Product Details