Product Details

SNP ID
rs199639375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34526240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTTAATCCTGCCTATCGGCCTC[A/T]CCTAACCTTGGTCTTGTTTTCTGGC
Phenotype
MIM: 607167
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DYNLRB1 PubMed Links

Gene Details

Gene
DYNLRB1
Gene Name
dynein light chain roadblock-type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281727.1 Intron NP_001268656.1
NM_001281728.1 Intron NP_001268657.1
NM_001281729.1 Intron NP_001268658.1
NM_001319157.1 Intron NP_001306086.1
NM_014183.3 Intron NP_054902.1
NM_177953.2 Intron NP_808852.1

View Full Product Details