Product Details

SNP ID
rs11546144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144106920 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGTGGAGGAAGAAAGCAGAAGC[A/G]GGGGCAGTGGGGCCGAGGAGACCAG
Phenotype
MIM: 123980 MIM: 610210 MIM: 611885
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYC1 PubMed Links

Gene Details

Gene
CYC1
Gene Name
cytochrome c1
There are no transcripts associated with this gene.

Gene
MAF1
Gene Name
MAF1 homolog, negative regulator of RNA polymerase III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032272.4 1330 Missense Mutation AGG,GGG R236G NP_115648.2
XM_017013903.1 1330 Missense Mutation AGG,GGG R236G XP_016869392.1
XM_017013904.1 1330 Intron XP_016869393.1
Gene
SHARPIN
Gene Name
SHANK associated RH domain interactor
There are no transcripts associated with this gene.

Gene
WDR97
Gene Name
WD repeat domain 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316309.1 1330 Intron NP_001303238.1
XM_017013385.1 1330 Intron XP_016868874.1
XM_017013386.1 1330 Intron XP_016868875.1
XM_017013387.1 1330 Intron XP_016868876.1
XM_017013388.1 1330 Intron XP_016868877.1
XM_017013389.1 1330 Intron XP_016868878.1
XM_017013390.1 1330 Intron XP_016868879.1
XM_017013391.1 1330 Intron XP_016868880.1
XM_017013392.1 1330 Intron XP_016868881.1
XM_017013393.1 1330 Intron XP_016868882.1
XM_017013394.1 1330 Intron XP_016868883.1

View Full Product Details