Product Details

SNP ID
rs5030686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52845630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAAACCCGGGGAGGCACCGTA[A/G]GGCTGCAGCAGGGAATAGGGGAATG
Phenotype
MIM: 603580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDH8 PubMed Links

Gene Details

Gene
PCDH8
Gene Name
protocadherin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002590.3 2838 Silent Mutation CCC,CCT P878P NP_002581.2
NM_032949.2 2838 Silent Mutation CCC,CCT P781P NP_116567.1

View Full Product Details