Product Details

SNP ID
rs539778490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:26947189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGAAGAGCCCGAGAGCCGGTAC[A/G]GAAGGGCGGCTGGGAAGGGGCGCGC
Phenotype
MIM: 608040 MIM: 603873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT74 PubMed Links

Gene Details

Gene
IFT74
Gene Name
intraflagellar transport 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099222.1 285 Intron NP_001092692.1
NM_001099223.1 285 Intron NP_001092693.1
NM_001099224.1 285 Intron NP_001092694.1
NM_025103.2 285 Intron NP_079379.2
XM_011518035.2 285 Intron XP_011516337.1
XM_011518036.2 285 Intron XP_011516338.1
XM_017015163.1 285 Intron XP_016870652.1
XM_017015164.1 285 Intron XP_016870653.1
Gene
IFT74-AS1
Gene Name
IFT74 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PLAA
Gene Name
phospholipase A2 activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031689.2 285 UTR 5 NP_001026859.1
NM_001321546.1 285 UTR 5 NP_001308475.1
XM_011518072.2 285 UTR 5 XP_011516374.1
XM_017015319.1 285 UTR 5 XP_016870808.1

View Full Product Details