Product Details
- SNP ID
-
hCV25762847
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:16744534 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAAAAAAACTTGAGAGTTACAGG[G/T]AGACTTTAGATTTCGTAATACTTAT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C11orf58
PubMed Links
Gene Details
- Gene
- C11orf58
- Gene Name
- chromosome 11 open reading frame 58
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014267.5 |
|
Intron |
|
|
NP_055082.1 |
View Full Product Details