Product Details

SNP ID
rs6442066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47414735 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGGAAATGCCCTCTGTTCTTCA[C/T]CAGGACTCTTCCAGCCTCTCCCTGA
Phenotype
MIM: 606584 MIM: 601510
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN23 PubMed Links

Gene Details

Gene
PTPN23
Gene Name
protein tyrosine phosphatase, non-receptor type 23
There are no transcripts associated with this gene.

Gene
SCAP
Gene Name
SREBF chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320044.1 Intron NP_001306973.1
NM_012235.3 Intron NP_036367.2
XM_005264967.1 Intron XP_005265024.1
XM_005264968.1 Intron XP_005265025.1
XM_005264971.1 Intron XP_005265028.1
XM_011533501.1 Intron XP_011531803.1
XM_011533502.2 Intron XP_011531804.1
XM_017005918.1 Intron XP_016861407.1
XM_017005919.1 Intron XP_016861408.1
XM_017005920.1 Intron XP_016861409.1
XM_017005921.1 Intron XP_016861410.1

View Full Product Details