Product Details

SNP ID
rs56351891
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:151265710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCAGGCTGGATAGTCCTAGACA[C/G]TACCAATCACTCATTTTCCAAGACC
Phenotype
MIM: 601648
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PSMD4 PubMed Links

Gene Details

Gene
PSMD4
Gene Name
proteasome 26S subunit, non-ATPase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002810.2 Intron NP_002801.1
XM_005245354.2 Intron XP_005245411.1
XM_006711464.3 Intron XP_006711527.1
XM_017001856.1 Intron XP_016857345.1

View Full Product Details