Product Details

SNP ID
hCV25766581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178466188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACTCGAAGATTAATATTTAAAC[G/T]GAATTTTGTTGAAACTAAATGCTTT
Phenotype
MIM: 610219 MIM: 607062
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DFNB59 PubMed Links

Gene Details

Gene
DFNB59
Gene Name
deafness, autosomal recessive 59
There are no transcripts associated with this gene.

Gene
FKBP7
Gene Name
FK506 binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135212.1 Intron NP_001128684.1
NM_181342.2 Intron NP_851939.1

View Full Product Details