Product Details

SNP ID
rs35075981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55009297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATTGCAGGCAGCCACAGACAC[A/G]TCCTGGAGGGGCAGCCCCACCGCAG
Phenotype
MIM: 301300 MIM: 300773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALAS2 PubMed Links

Gene Details

Gene
ALAS2
Gene Name
5'-aminolevulinate synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000032.4 1781 Silent Mutation GAC,GAT D549D NP_000023.2
NM_001037967.3 1781 Silent Mutation GAC,GAT D512D NP_001033056.1
NM_001037968.3 1781 Silent Mutation GAC,GAT D536D NP_001033057.1
XM_005261995.3 1781 Silent Mutation GAC,GAT D573D XP_005262052.1
XM_011530771.1 1781 Silent Mutation GAC,GAT D262D XP_011529073.1
XM_017029354.1 1781 Intron XP_016884843.1
Gene
APEX2
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 2
There are no transcripts associated with this gene.

View Full Product Details