Product Details
- SNP ID
-
rs16866373
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:178527353 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAAGGTCTTAGAATCACTGAAAA[A/T]AATAAAGATTTGCTTTCTACTGAAT
- Phenotype
-
MIM: 188840
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TTN
PubMed Links
Gene Details
- Gene
- TTN
- Gene Name
- titin
- Gene
- TTN-AS1
- Gene Name
- TTN antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details