Product Details

SNP ID
rs115969402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81458492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAACAGGCTAAACTTGCCCTGCC[C/T]CTTTCTCCTCAGACCAAACCCTTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FABP9 PubMed Links

Gene Details

Gene
FABP9
Gene Name
fatty acid binding protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080526.1 Intron NP_001073995.1

View Full Product Details