Product Details

SNP ID
rs35954676
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:151171484 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTACCAGACTGAAGCTCCGCAC[A/G]TAGGTATTTGCCTTCATTGCCTCAC
Phenotype
MIM: 608095 MIM: 605834 MIM: 600607
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYSMD1 PubMed Links

Gene Details

Gene
LYSMD1
Gene Name
LysM domain containing 1
There are no transcripts associated with this gene.

Gene
SCNM1
Gene Name
sodium channel modifier 1
There are no transcripts associated with this gene.

Gene
TMOD4
Gene Name
tropomodulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013353.2 747 Silent Mutation TAA,TAG *225* NP_037485.2
XM_011509449.1 747 Nonsense Mutation TAA,TAG *225* XP_011507751.1
XM_017001089.1 747 Nonsense Mutation TAA,TAG *225* XP_016856578.1
XM_017001090.1 747 Intron XP_016856579.1
Gene
TNFAIP8L2-SCNM1
Gene Name
TNFAIP8L2-SCNM1 readthrough
There are no transcripts associated with this gene.

Gene
VPS72
Gene Name
vacuolar protein sorting 72 homolog
There are no transcripts associated with this gene.

View Full Product Details