Product Details

SNP ID
rs28730722
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:151171494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAGCTCCGCACATAGGTATTT[C/G]CCTTCATTGCCTCACACAGCTCACT
Phenotype
MIM: 608095 MIM: 605834 MIM: 600607
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYSMD1 PubMed Links

Gene Details

Gene
LYSMD1
Gene Name
LysM domain containing 1
There are no transcripts associated with this gene.

Gene
SCNM1
Gene Name
sodium channel modifier 1
There are no transcripts associated with this gene.

Gene
TMOD4
Gene Name
tropomodulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013353.2 737 Missense Mutation GCA,GGA A222G NP_037485.2
XM_011509449.1 737 Missense Mutation GCA,GGA A222G XP_011507751.1
XM_017001089.1 737 Missense Mutation GCA,GGA A222G XP_016856578.1
XM_017001090.1 737 Intron XP_016856579.1
Gene
TNFAIP8L2-SCNM1
Gene Name
TNFAIP8L2-SCNM1 readthrough
There are no transcripts associated with this gene.

Gene
VPS72
Gene Name
vacuolar protein sorting 72 homolog
There are no transcripts associated with this gene.

View Full Product Details