Product Details
- SNP ID
-
rs57185523
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149432989 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GCTGCTGCCTGGCGCCCCTAACCTA[C/T]CTGGATGGAGGTACAGCACTGCTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF777
PubMed Links
Gene Details
- Gene
- ZNF777
- Gene Name
- zinc finger protein 777
View Full Product Details