Product Details

SNP ID
rs61731829
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:200873415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGGCGGGCACGCGCTGCGCGGG[A/C]GCGCTGCTGCTGGCGGGCATGAGCG
Phenotype
MIM: 602174
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR25 PubMed Links

Gene Details

Gene
GPR25
Gene Name
G protein-coupled receptor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005298.3 461 Silent Mutation GGA,GGC G126G NP_005289.2

View Full Product Details