Product Details

SNP ID
rs138412526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118036582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACTCAAACGGAGGGAGGGAAAG[A/G]TACCACCACCAAATCTGCAGGCCCT
Phenotype
MIM: 600407
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RFC5 PubMed Links

Gene Details

Gene
RFC5
Gene Name
replication factor C subunit 5
There are no transcripts associated with this gene.

Gene
WSB2
Gene Name
WD repeat and SOCS box containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278557.1 Intron NP_001265486.1
NM_001278558.1 Intron NP_001265487.1
NM_018639.4 Intron NP_061109.1
XM_017019642.1 Intron XP_016875131.1

View Full Product Details