Product Details

SNP ID
rs61748963
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:247605760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGATCCCCCTCTTCATACCCCA[A/G]TGTACTTTTTTCTCAGCAACCTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2G3 PubMed Links

Gene Details

Gene
OR2G3
Gene Name
olfactory receptor family 2 subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001914.1 175 Missense Mutation ATG,GTG M59V NP_001001914.1

View Full Product Details