Product Details

SNP ID
rs3798964
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:161351917 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAGAGGAAAAGTCCCTTATCTTGG[A/G]ACTTATTTTGGAATTATTCACCTCC
Phenotype
MIM: 602544
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARK2 PubMed Links

Gene Details

Gene
PARK2
Gene Name
parkin RBR E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004562.2 Intron NP_004553.2
NM_013987.2 Intron NP_054642.2
NM_013988.2 Intron NP_054643.2
XM_011535863.1 Intron XP_011534165.1
XM_017010908.1 Intron XP_016866397.1
XM_017010909.1 Intron XP_016866398.1

View Full Product Details