Product Details
- SNP ID
-
rs3768339
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:35184401 on Build GRCh38
- Set Membership
-
JSNP
Validated
- Context Sequence [VIC/FAM]
- TACTAAAATGCAAGAATTTAAAAGA[C/T]TGGTATCTAAACAAAAAAACAAAAC
- Phenotype
-
MIM: 605199
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SFPQ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72908970] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SFPQ
- Gene Name
- splicing factor proline and glutamine rich
View Full Product Details