Product Details

SNP ID
rs34353469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:118924469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTACATTGTGAAGCTGTGAAGGA[A/C]CCTGATGGTCTTCTTTAGACTCTGA
Phenotype
MIM: 602643
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TNFRSF11B PubMed Links

Gene Details

Gene
TNFRSF11B
Gene Name
TNF receptor superfamily member 11b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002546.3 1434 Missense Mutation GTC,TTC V371F NP_002537.3

View Full Product Details