Product Details

SNP ID
rs60185051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13212034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGGGATGCACTGGGCTGCTTG[G/T]GGGGGGGCCTGGCCCTACCCAGTGC
Phenotype
MIM: 601011
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CACNA1A PubMed Links

Gene Details

Gene
CACNA1A
Gene Name
calcium voltage-gated channel subunit alpha1 A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000068.3 Intron NP_000059.3
NM_001127221.1 Intron NP_001120693.1
NM_001127222.1 Intron NP_001120694.1
NM_001174080.1 Intron NP_001167551.1
NM_023035.2 Intron NP_075461.2

View Full Product Details