Product Details

SNP ID
rs34079256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9023004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCACCGACTCGCACACCGACATG[A/C]GCCCGCTCTCGGGCGGCCTCATCGC
Phenotype
MIM: 601614
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NTN1 PubMed Links

Gene Details

Gene
NTN1
Gene Name
netrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004822.2 838 Missense Mutation AGC,CGC S211R NP_004813.2
XM_006721595.3 838 Missense Mutation AGC,CGC S211R XP_006721658.1

View Full Product Details