Product Details

SNP ID
rs10252253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144110211 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTGTTTGTCTTAGTCGGGCCTC[C/T]TTCCTTGATTCTGGTCTCCTACATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2A2 PubMed Links

Gene Details

Gene
OR2A2
Gene Name
olfactory receptor family 2 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005480.2 629 Missense Mutation CCT,CTT P210L NP_001005480.2

View Full Product Details