Product Details

SNP ID
rs17122157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118405777 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCAGAAATACTCAAATGGAGGCC[A/G]TTAATATGAGGAAAACACCAAGAGC
Phenotype
MIM: 603753
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5L PubMed Links

Gene Details

Gene
ATP5L
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006476.4 Intron NP_006467.4
Gene
LOC100131626
Gene Name
uncharacterized LOC100131626
There are no transcripts associated with this gene.

Gene
UBE4A
Gene Name
ubiquitination factor E4A
There are no transcripts associated with this gene.

View Full Product Details