Product Details

SNP ID
rs61732639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186930832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCTCCGCGGCCGAGAGTGCAG[A/C]GAGTGAGTAGGGGGTGCGGTGGGCG
Phenotype
MIM: 109675
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ST6GAL1 PubMed Links

Gene Details

Gene
ST6GAL1
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003032.2 490 Intron NP_003023.1
NM_173216.2 490 UTR 5 NP_775323.1
NM_173217.2 490 UTR 5 NP_775324.1
XM_005247719.1 490 Intron XP_005247776.1
XM_005247720.1 490 Intron XP_005247777.1
XM_006713734.1 490 UTR 5 XP_006713797.1
XM_011513085.1 490 UTR 5 XP_011511387.1
XM_011513086.1 490 Intron XP_011511388.1
XM_017007065.1 490 UTR 5 XP_016862554.1
XM_017007066.1 490 Intron XP_016862555.1
XM_017007067.1 490 Intron XP_016862556.1
XM_017007068.1 490 Intron XP_016862557.1

View Full Product Details