Product Details

SNP ID
rs111521556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57546961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGGCCTTGCTGTATTCATTGCA[A/G]CAATAGGGTTCTCTCATGTATGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF549 PubMed Links

Gene Details

Gene
ZNF549
Gene Name
zinc finger protein 549
There are no transcripts associated with this gene.

Gene
ZNF550
Gene Name
zinc finger protein 550
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277090.1 1461 UTR 3 NP_001264019.1
NM_001277091.1 1461 UTR 3 NP_001264020.1
NM_001277092.1 1461 UTR 3 NP_001264021.1
NM_001277093.1 1461 Intron NP_001264022.1
XM_011526567.2 1461 UTR 3 XP_011524869.1
XM_011526568.2 1461 UTR 3 XP_011524870.1
XM_017026401.1 1461 UTR 3 XP_016881890.1

View Full Product Details