Product Details

SNP ID
rs144032498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57647397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACCCATACCTCTACTTTCCCC[A/G]TCACCATCGACCCCCCCACAGCATC
Phenotype
MIM: 610175 MIM: 609725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR130A PubMed Links

Gene Details

Gene
MIR130A
Gene Name
microRNA 130a
There are no transcripts associated with this gene.

Gene
YPEL4
Gene Name
yippee like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145008.2 Intron NP_659445.1
XM_017017318.1 Intron XP_016872807.1

View Full Product Details