Product Details

SNP ID
rs201382755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200871564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACATATGAAAATTTCCTTAAATT[A/G]AAACATGATATCCATATCTATGACA
Phenotype
MIM: 601951 MIM: 615811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLK1 PubMed Links

Gene Details

Gene
CLK1
Gene Name
CDC like kinase 1
There are no transcripts associated with this gene.

Gene
PPIL3
Gene Name
peptidylprolyl isomerase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032472.3 Intron NP_115861.1
NM_130906.2 Intron NP_570981.1
XM_005246651.3 Intron XP_005246708.1
XM_005246652.4 Intron XP_005246709.1
XM_011511358.2 Intron XP_011509660.1
XM_017004352.1 Intron XP_016859841.1
XM_017004353.1 Intron XP_016859842.1
XM_017004354.1 Intron XP_016859843.1
XM_017004355.1 Intron XP_016859844.1
XM_017004356.1 Intron XP_016859845.1
XM_017004357.1 Intron XP_016859846.1
XM_017004358.1 Intron XP_016859847.1
XM_017004359.1 Intron XP_016859848.1
XM_017004360.1 Intron XP_016859849.1

View Full Product Details